Learn More
Sino Biological Pan Influenza B Nucleoprotein Antibody, Rabbit MAb
Supplier:  Sino Biological 40438R00420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza B (B/Florida/4/2006) Nucleoprotein / NP Protein (Catalog# 40438-V08B; ACF54251.1; Met1-Tyr560).
Catalog No. 50-237-4354