Learn More
Sino Biological Influenza B (B/Brisbane/60/2008) Nucleoprotein / NP Protein (His Tag)
Supplier:  Sino Biological 40783V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Brisbane/60/2008) nucleoprotein (translated amino acids of EPI902986) (Met1-Tyr560) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3588