Learn More
Creative Biomart Recombinant HAV VP1-P2A Antigen, GST-tagged
Supplier:  Creative Biomart VAC0204
Encompass_Preferred
The E. coli derived 51.2 kDa recombinant protein fused to a GST tag contains the VP1-P2A immunodominant regions, amino acids 722-830.
Catalog No. 50-208-5044
 Restricted Item