Learn More
Creative Biomart Recombinant HAV VP4-VP2 Antigen, GST-tagged
Supplier:  Creative Biomart VAC0205
Encompass_Preferred
The E. coli derived 44 kDa recombinant protein fused to a GST tag contains the VP4-VP2 immunodominant regions, amino acids 55-164.
Catalog No. 50-208-5045