Learn More
Creative Biomart Recombinant HAV P2C Antigen
Supplier:  Creative Biomart VAC0200
Encompass_Preferred
The E. coli derived recombinant 38.5 kDa protein contains the P2C immunodominant regions, amino acids 1121-1234.
Catalog No. 50-208-5040