Learn More
Creative Biomart Recombinant HAV P2C-P3A Antigen
Supplier:  Creative Biomart VAC0201
Encompass_Preferred
The E. coli derived recombinant protein contains the P2C-P3A immunodominant regions, amino acids 1392-1521.
Catalog No. 50-208-5041