Learn More
Creative Biomart Recombinant EBV Early Antigen Type D, GST-tagged
Catalog No. 502085036
Encompass_Preferred
The E. coli derived 35.5 kDa recombinant protein contains the HHV-4 Early Antigen Type D, C-terminus regions amino acids 306-390. The protein is fused to a 26 kDa GST tag.
Catalog No. 50-208-5036 Supplier Creative Biomart Supplier No. VAC0196
 Restricted Item
Add to Cart
Add to Cart
The E. coli derived 35.5 kDa recombinant protein contains the HHV-4 Early Antigen Type D, C-terminus regions amino acids 306-390. The protein is fused to a 26 kDa GST tag.