Learn More
Creative Biomart Recombinant Ebola Zaire Nucleoprotein Antigen, His-tagged
Catalog No. 502085034
Encompass_Preferred
Nucleoprotein (NP) of Ebola virus has strong antigenicity in immune reactions. C-terminal of EBO-Z nucleoprotein was expressed and purified from E. coli, it migrated at 15kDa on SDS-PAGE and pI is 4.8
Catalog No. 50-208-5034 Supplier Creative Biomart Supplier No. VAC0194
Nucleoprotein (NP) of Ebola virus has strong antigenicity in immune reactions. C-terminal of EBO-Z nucleoprotein was expressed and purified from E. coli, it migrated at 15kDa on SDS-PAGE and pI is 4.8