Learn More
Creative Biomart Influenza B virus (Tokio) Antigen
Supplier:  Creative Biomart VAC0159
Encompass_Preferred
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza B virus, strain B/Tokio/53/99. The Influenza B Virus was purified by Ultracentrifugation with 10-40 % sucrose gradient.
Catalog No. 50-208-4999