Learn More
Creative Biomart Recombinant CMV gB Antigen, GST-tagged
Supplier:  Creative Biomart VAC0186
Encompass_Preferred
The E. coli derived recombinant artificial mosaic protein contains the CMV gB immunodominant regions 11-67 amino acids, fused with a 26 kDa GST Tag, the protein weight is 6.5 kDa, having a total weigh
Catalog No. 50-208-5026