Learn More
Creative Biomart Recombinant Chlamydia W4-W5 Antigen, His-tagged
Supplier:  Creative Biomart VAC0183
Encompass_Preferred
The E. coli derived recombinant 6 His fusion at C-terminus protein contains Chlamydia Trachomatis MOMP protein epitopes, 191-354 amino acids.
Catalog No. 50-208-5023