Learn More
Creative Biomart Recombinant Chlamydia W2 Antigen, His-tagged
Supplier:  Creative Biomart VAC0180
Encompass_Preferred
The E. coli derived recombinant 6 His fusion at C-terminus protein contains Chlamydia Trachomatis MOMP protein epitopes, 66-165 amino acids.
Catalog No. 50-208-5020