Learn More
Creative Biomart Recombinant Chlamydia W3-W6 Antigen, His-tagged
Supplier:  Creative Biomart VAC0181
Encompass_Preferred
The E. coli derived recombinant 6 His fusion at C-terminus. protein contains Chlamydia Trachomatis MOMP protein epitopes, 128-398 amino acids.
Catalog No. 50-208-5021
 Restricted Item