Learn More
Creative Biomart Recombinant CHIKV E1 Antigen
Supplier:  Creative Biomart VAC0175
Encompass_Preferred
Recombinant Chikungunya Wild Type E1 produced in insect cells is a polypeptide chain containing amino acids 1-415 and having a molecular weight of approximately 50 kDa.CHIKV E1 is purified by propriet
Catalog No. 50-208-5015
 Restricted Item