Learn More
ATCC Influenza B virus; B/GL/1739/54
Catalog No. 502382927
Encompass
Influenza B virus strain B/GL/1739/54 is propagated in SPF chicken embryonated eggs. This whole-genome sequenced strain was isolated from a human patient at Great Lakes Naval Training Station in Illinois in 1954 and was deposited by the Centers for Disease Control and Prevention. Influenza B virus is used in respiratory disease research and assay development.
Catalog No. 50-238-2927 Supplier ATCC Supplier No. VR103
Influenza B virus strain B/GL/1739/54 is propagated in SPF chicken embryonated eggs. This whole-genome sequenced strain was isolated from a human patient at Great Lakes Naval Training Station in Illinois in 1954 and was deposited by the Centers for Disease Control and Prevention. Influenza B virus is used in respiratory disease research and assay development.