Learn More
ATCC Influenza B virus; B/Lee/40; TC adapted
Catalog No. 502382711
Encompass
Influenza B virus strain B/Lee/40 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated in 1940 from throat washings of an influenza patient in Irvington, New York, and was deposited by the Centers for Disease Control and Prevention. It has applications in respiratory disease research.
Catalog No. 50-238-2711 Supplier ATCC Supplier No. VR1535
Influenza B virus strain B/Lee/40 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated in 1940 from throat washings of an influenza patient in Irvington, New York, and was deposited by the Centers for Disease Control and Prevention. It has applications in respiratory disease research.