Learn More
ATCC Influenza B virus; B/Maryland/1/59
Supplier:  ATCC VR296
Encompass
Influenza B virus strain B/Maryland/1/59 is propagated in SPF embryonated chicken eggs. This strain was isolated in 1959 from a human in Maryland and was deposited by the Centers for Disease Control and Prevention. It has applications in influenza and respiratory research.
Catalog No. 50-238-4369
 Restricted Item