Learn More
ATCC Influenza B virus; B/Taiwan/2/62; RNA
Catalog No. 502382265
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from MDCK cells (ATCC CCL-34) infected with Influenza B virus strain B/Taiwan/2/62 (ATCC VR-1735). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other molecular based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-2265 Supplier ATCC Supplier No. VR1735D
Genomic RNA isolated from a preparation of cell lysate and supernatant from MDCK cells (ATCC CCL-34) infected with Influenza B virus strain B/Taiwan/2/62 (ATCC VR-1735). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other molecular based procedures. The source organism and host cells are also available through the ATCC catalog.