Learn More
ATCC Influenza B virus; B/Taiwan/2/62; TC adapted
Catalog No. 502382264
Encompass
Influenza B virus strain B/Taiwan/2/62 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated in 1962 from a human patient in Taiwan and was deposited by the Centers for Disease Control and Prevention. It has applications in respiratory and influenza research.
Catalog No. 50-238-2264 Supplier ATCC Supplier No. VR1735
Influenza B virus strain B/Taiwan/2/62 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated in 1962 from a human patient in Taiwan and was deposited by the Centers for Disease Control and Prevention. It has applications in respiratory and influenza research.