Learn More
ATCC Influenza B virus; B/Taiwan/2/62
Catalog No. 502381682
Encompass
Influenza B virus strain B/Taiwan/2/62 is propagated in SPF chicken embryonated eggs. This strain was isolated in 1962 from a human patient in Taiwan and was deposited by the Centers for Disease Control and Prevention. It has applications in influenza research.
Catalog No. 50-238-1682 Supplier ATCC Supplier No. VR295
Influenza B virus strain B/Taiwan/2/62 is propagated in SPF chicken embryonated eggs. This strain was isolated in 1962 from a human patient in Taiwan and was deposited by the Centers for Disease Control and Prevention. It has applications in influenza research.