Learn More
ATCC Influenza B virus; B/Wisconsin/1/2010
Supplier:  ATCC VR1883
Encompass
Influenza B virus (Yamagata Lineage) strain B/Wisconsin/1/2010 is propagated in SPF embryonated chicken eggs. The virus was isolated in 2010 from a human patient in Wisconsin. It has applications in infectious disease and respiratory disease research and vaccine development.
Catalog No. 50-238-4770