Learn More
ATCC Influenza B virus (BY); B/Massachusetts/2/2012
Catalog No. 502384772
Encompass
Influenza B virus strain B/Massachusetts/2/2012 is propagated in SPF chicken embryonated eggs. This strain was isolated in 2012 from a human in Massachusetts and was deposited by Massachusetts Department of Public Health. This virus is whole-genome sequenced and has applications in vaccine development and influenza research.
Catalog No. 50-238-4772 Supplier ATCC Supplier No. VR1813
Influenza B virus strain B/Massachusetts/2/2012 is propagated in SPF chicken embryonated eggs. This strain was isolated in 2012 from a human in Massachusetts and was deposited by Massachusetts Department of Public Health. This virus is whole-genome sequenced and has applications in vaccine development and influenza research.