Learn More
ATCC Influenza B virus (BY); RNA; B/Massachusetts/2/2012
Catalog No. 502381979
Encompass
Genomic RNA isolated from a preparation of Influenza B virus (BY) strain B/Massachusetts/2/2012 (ATCC VR-1813) propagated in SPF chicken embryonated eggs. This product was prepared using methods known to inactivate viruses. The source organism is also available through the ATCC catalog.
Catalog No. 50-238-1979 Supplier ATCC Supplier No. VR1813D
Add to Cart
Add to Cart
Genomic RNA isolated from a preparation of Influenza B virus (BY) strain B/Massachusetts/2/2012 (ATCC VR-1813) propagated in SPF chicken embryonated eggs. This product was prepared using methods known to inactivate viruses. The source organism is also available through the ATCC catalog.