Learn More
Zeptometrix Influenza B Virus (Strain: Washington/02/19) Culture Fluid
SDP
Catalog No. 22158419 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-158-419 1 mL
1 options
Catalog No. 22-158-419 Supplier Zeptometrix Supplier No. 0810611CF
Add to Cart
Add to Cart
This product is intended for research, analytical testing during product development, or quality assurance testing. Viral culture fluids are sold as consumable testing materials, and are not for propagation or commercialization.
Influenza B Virus (Strain: Washington/02/19) is an enveloped virus with a diameter of 80 to 120 nm, and contains a single-stranded, segmented, negative-sense RNA within a nucleocapsid. Each frozen aliquot contains 1 mL of titered viral culture fluid.
Specifications
Biosafety Level 2
Certifications/Compliance ISO 13485
Product Type Infectous Culture Fluid
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Quantity 1 mL
Regulatory Status RUO
The purchase of infectious microorganisms from ZeptoMetrix requires a Material Transfer Agreement (MTA).