Learn More
Bioworld Influenza B Virus Victoria 504/00 (Recombinant), 10 μg
Supplier:  Bioworld 220605231
Encompass_Preferred
Influenza virus, type B Victoria 504/00 (InfluVictoria) Source: Allantoic fluid Purification: Purified by ultracentrifugation using 10-40% sucrose gradient. Presentation: Solution (1.1mg/1ml) in STE containing 0.1% NaN3 and 0.005% merthiolate. Inactivation: 0.005% merthiolate This product has been treated in a manner consistent with methods of inactivation. Generally accepted good laboratory practices appropriate to microbiological/viral safe handling practices and techniques is required at work. Analysis: Purity more than 90% according to SDS-PAGE. Immunological activity: Has been tested with anti-influenza B monoclonal antibodies in ELISA. Application: Serological studies of influenza B virus, immunogen for antibody production. Storage: -20 C. Usage: BioWORLD's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-621