Learn More
Zeptometrix Influenza D Virus (Bovine/Mississippi/C00046N/14) Culture Fluid
SDP
Catalog No. 22158407 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-158-407 1 mL
1 options
Catalog No. 22-158-407 Supplier Zeptometrix Supplier No. 0810567CF
Add to Cart
Add to Cart
Influenza D Virus (Strain: Bovine/Mississippi/C00046N/14) is an enveloped virus with a diameter of 80 to 120 nm, and contains a single stranded, segmented, negative-sense RNA within a nucleocapsid.
Viral culture fluids are sold as consumable testing materials; propagation or commercialization is prohibited without prior written consent from ZeptoMetrix. The suitability and performance characteristics should be determined by your laboratory for each intended usage. These products are NOT intended for use in the manufacture or processing of injectable products subject to licensure under section 351 of the Public Health Service Act or for any other product intended for administration to humans.
Specifications
Type Culture Fluid
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Quantity 1 mL
The purchase of infectious microorganisms from ZeptoMetrix requires a Material Transfer Agreement (MTA).