Learn More
Abcam Human Anti-Influenza virus A IgG ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1087451X96TESTS
Encompass_Preferred
Human Anti-Influenza virus A IgG ELISA Kit is a Indirect ELISA kit for the measurement of Human Anti-Influenza virus A IgG in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1482069