Learn More
Sino Biological Influenza HA ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological SEK1168415
Encompass_Preferred
Each vial contains 144 ng of recombinant H1N1 (A/Puerto Rico/8/1934) HA. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 4000 pg/mL is recommended.
Catalog No. 50-237-3433