Learn More
Sino Biological Influenza HA ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502371570
Encompass_Preferred
Each vial contains 1050 ng of recombinant Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin / HA. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 25000 pg/mL is recommended.
Catalog No. 50-237-1570 Supplier Sino Biological Supplier No. SEK1105615
Each vial contains 1050 ng of recombinant Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin / HA. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 25000 pg/mL is recommended.