Learn More
Sino Biological Influenza NP PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11675T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A Nucleoprotein / NP ( Catalog#11675-V08B; AAM75159.1;Met1-Gly490, Ile 116 Met). Influenza A Nucleoprotein / NP specific IgG was purified by Influenza A Nucleoprotein / NP affinity chromatography.
Catalog No. 50-237-1628