Learn More
Description
- Composed of 20 members
- Eight vials Influenza A/B Non-Reactive (0.5mL each), Seven vials Influenza A Reactive (0.5mL each) and Five vials Influenza B Reactive (0.5mL each)
- Prepared from inactivated viral culture fluids
- Processed to eliminate contaminants
- Needs evaluation of members as per instructions
Not substitute for mandatory positive and negative control reagents provided with licensed test kits.
For training, lot-to-lot comparison of reagent test kits and to evaluate and compare intra laboratory and inter laboratory performance of Influenza testing platforms
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Product Type | Validation Panel |
| For Use With (Application) | In vitro assay procedures |
| Quantity | 20 x 0.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
