Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ BinaxNOW™ Influenza A&B Card 2 Swab Kit (10-Pos/Neg Swabs/Kit)

Catalog No. 23046689 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-689 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-046-689 Supplier Abbott™ Supplier No. 575080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Influenza testing

  • Test card
  • For influenza testing

Specifications

Content And Storage Room Temperature
Description BinaxNOW™ Influenza A and B Card 2 Swab Kit
CE Marker No
Format Swab
Sample Type Swab
Type Swab
Formulation Positive and negative control swabs
For Use With (Application) Influenza testing
Includes 10 positive and 10 negative swabs
Packaging Type Individually Wrapped 20/box
Quantity 20
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.