Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Calibration Check Card Pack (2 per pack)

Catalog No. 23046685 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-685 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-685 Supplier Abbott™ Supplier No. LFR003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in testing influenza

  • Calibration check card
  • For influenza testing

Specifications

Content And Storage Room Temperature
Description Calibration Check Card Pack
Format Card
Sample Type Card
Type Card
For Use With (Application) Influenza testing
Includes 2 calibration check cards
Quantity 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.