Learn More
Abbott™ DIGIVAL™ Complete Kit
Catalog No. 23046680 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
For Use With (Application):
Influenza testing
Catalog No. For Use With (Application)
23-046-680 Influenza testing
1 options
This item is not returnable. View return policy
Catalog No. 23-046-680 Supplier Abbott™ Supplier No. LFR124
Add to Cart
Add to Cart
This item is not returnable. View return policy
DIGIVAL™ Complete Kit is an in vitro immunochromatographic assay for the qualitative detection of influenza A and B nucleoprotein antigens in nasopharyngeal swabs and nasal swab samples.
  • Includes: DIGIVAL, barcode scanner, USB, calibration check card pack, and tray
  • For use with the BinaxNOW™ Influenza A&B Card 2
Specifications
Description Reader
For Use With (Application) Influenza testing
Includes Reader, barcode scanner, calibration check card and tray, USB