Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ BinaxNOW™ Influenza A&B Card 2 (22 Test)

Catalog No. 23046682 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
22
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-682 22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-046-682 Supplier Abbott™ Supplier No. 575000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with influenza testing.

  • Test card
  • For influenza testing

Specifications

Content And Storage Room Temperature
Description BinaxNOW™ Influenza A and B Card 2
CE Marker No
Clia Complexity Waived
Detectable Analytes Influenza
Format Card
Sample Type Nasal and NP
Type Lateral Flow Assay
Formulation LF test card
For Use With (Application) Influenza testing
Includes Controls, collection swabs
Packaging Type Box
Quantity 22
Test Time 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.