Learn More
Cayman Chemical InfluenzaAH1N1SwInFlu220uL
SDP
Catalog No. 502919145
Encompass_Preferred
An active metabolite of thalidomide and a PROTAC synthetic intermediate decreases production of TNF-a induced by TPA in THP-1 cells at 30 UM completely inhibits the proliferation HUVECs at 333 UM inhibits VEGF- and FGF2-induced angiogenesis by 14 in a CAM assay in utero at 100 Ug/embryo
Catalog No. 50-291-9145 Supplier Cayman Chemical Supplier No. 3954120 UL
An active metabolite of thalidomide and a PROTAC synthetic intermediate decreases production of TNF-a induced by TPA in THP-1 cells at 30 UM completely inhibits the proliferation HUVECs at 333 UM inhibits VEGF- and FGF2-induced angiogenesis by 14 in a CAM assay in utero at 100 Ug/embryo