Learn More
Cayman Chemical InfluenzaH1N1 HA MoncLNa 50uL
SDP
Catalog No. 502915864
Encompass_Preferred
An activator of KLF4 increases levels of KLF4 and induces apoptosis in HCC1187 HCC38 and MDA-MB-468 cancer cells at 5 and 10 UM cytotoxic to a panel of 16 AML and lymphoma cell lines (IC50s 0.057-1.75 UM) induces cell cycle arrest at the G0/G1 phase in MV4-11 KG-1 and EoL-1 cells accelerates wound healing as well as increases plasma levels of KLF4 and wound site MDSC accumulation in an ob/ob mouse model of diabetic wound healing
Catalog No. 50-291-5864 Supplier Cayman Chemical Supplier No. 3810450 UL
An activator of KLF4 increases levels of KLF4 and induces apoptosis in HCC1187 HCC38 and MDA-MB-468 cancer cells at 5 and 10 UM cytotoxic to a panel of 16 AML and lymphoma cell lines (IC50s 0.057-1.75 UM) induces cell cycle arrest at the G0/G1 phase in MV4-11 KG-1 and EoL-1 cells accelerates wound healing as well as increases plasma levels of KLF4 and wound site MDSC accumulation in an ob/ob mouse model of diabetic wound healing