Learn More
Electron Microscopy Sciences INFINITY3-3UR 2.8 Megapixel USB 3.0 Microscopy Camera, Uncooled CCD, Color
SDP
Catalog No. 501927235
Encompass_Preferred
The INFINITY3-3UR uncooled scientific digital camera features a Sony ICX674 CCD sensor. Designed for use in scientific and industrial applications requiring optimal color reproduction, extreme sensitivity, increased resolution and high speed.
Relying on Sony's EXview HAD ll technology, this camera offers extremely high dynamic range, 4.54x4.54 um pixels and very low noise.
INFINITY CAPTURE, an intuitive image capture program, and INFINITY ANALYZE, a full image analysis package offering camera control, measurement, annotation, tiling and post capture enhancement, are included with the camera.
53 fps at 2.8 megapixel, 66 fps at full HD (1936x1456).
High-speed USB 3.0 interface for ease of installation on any computer.
128 MB of onboard memory for frame buffering results in reliable image delivery for demanding systems.
Select 8 and 14-bit pixel data modes.
Sensor: Uncooled CCD
Type: Color
Catalog No. 50-192-7235 Supplier Electron Microscopy Sciences Supplier No. 95123
Add to Cart
Add to Cart
Request a Quote
The INFINITY3-3UR uncooled scientific digital camera features a Sony ICX674 CCD sensor. Designed for use in scientific and industrial applications requiring optimal color reproduction, extreme sensitivity, increased resolution and high speed.
Relying on Sony's EXview HAD ll technology, this camera offers extremely high dynamic range, 4.54x4.54 um pixels and very low noise.
INFINITY CAPTURE, an intuitive image capture program, and INFINITY ANALYZE, a full image analysis package offering camera control, measurement, annotation, tiling and post capture enhancement, are included with the camera.
53 fps at 2.8 megapixel, 66 fps at full HD (1936x1456).
High-speed USB 3.0 interface for ease of installation on any computer.
128 MB of onboard memory for frame buffering results in reliable image delivery for demanding systems.
Select 8 and 14-bit pixel data modes.
Sensor: Uncooled CCD
Type: Color