Learn More
Electron Microscopy Sciences INFINITY 3-6URC Uncooled CCD Color Camera
SDP
Catalog No. 501927440
Encompass_Preferred
The INFINITY3-6UR is a high-speed, high-sensitivity microscope camera with 6 megapixel resolution. With this high resolution and excellent color reproduction, speed and light sensitivity needed for low-light applications, it is ideal for microscopy applications. The camera incorporates Sony's ICX694 CCD sensor with high QE, high dynamic range and high speed. Full resolution 6 MP images are sent to a host computer at up to 27 frames per second (fps) over the plug and play USB 3.0 connection. The 'R' grade designation of the INFINITY3-6UR is a testament to low-noise electronics, high-grade components and unique thermal management techniques.
Catalog No. 50-192-7440 Supplier Electron Microscopy Sciences Supplier No. 95131
Add to Cart
Add to Cart
Request a Quote
The INFINITY3-6UR is a high-speed, high-sensitivity microscope camera with 6 megapixel resolution. With this high resolution and excellent color reproduction, speed and light sensitivity needed for low-light applications, it is ideal for microscopy applications. The camera incorporates Sony's ICX694 CCD sensor with high QE, high dynamic range and high speed. Full resolution 6 MP images are sent to a host computer at up to 27 frames per second (fps) over the plug and play USB 3.0 connection. The 'R' grade designation of the INFINITY3-6UR is a testament to low-noise electronics, high-grade components and unique thermal management techniques.