Learn More
Grainger Lovibond Infrared Interface Module, IRiM, 1 ea
Catalog No. 502756829
Encompass_Preferred
The Infrared Interface Module (IRiM) is a versatile middleware device designed to receive data via an infrared interface from compatible water testing instruments, such as photometers, and transmit that data to a PC, USB printer, or serial printer. It facilitates seamless data integration by converting infrared signals into formats compatible with common peripheral interfaces, supporting streamlined data acquisition and documentation.
  • Enables data transmission to USB PC interfaces, USB printers (specifically HP Deskjet 6940), or RS232 serial devices
  • Features user-selectable interface modes including PC, USB printer, and serial printer
  • Powered via USB connection or 4 AA batteries
  • Includes indicator LEDs for power, PC connection status, and interface selection
  • Requires Data Acquisition Software for PC-based data retrieval
Catalog No. 50-275-6829 Supplier Grainger Supplier No. 15F849
The Infrared Interface Module (IRiM) is a versatile middleware device designed to receive data via an infrared interface from compatible water testing instruments, such as photometers, and transmit that data to a PC, USB printer, or serial printer. It facilitates seamless data integration by converting infrared signals into formats compatible with common peripheral interfaces, supporting streamlined data acquisition and documentation.
  • Enables data transmission to USB PC interfaces, USB printers (specifically HP Deskjet 6940), or RS232 serial devices
  • Features user-selectable interface modes including PC, USB printer, and serial printer
  • Powered via USB connection or 4 AA batteries
  • Includes indicator LEDs for power, PC connection status, and interface selection
  • Requires Data Acquisition Software for PC-based data retrieval