Learn More
Andwin Scientific INFRARED LAMP
SDP
Catalog No. NC9423184
Encompass_Preferred
The Infrared Heating and Drying Lamp gives onvenient and dependable source for near-infrared rays for heating and drying purposes. The arm is offset in order to permit the evaporating dish or other object to be placed and heated directly under the lamp. The switch to turn it on or off is located on the lamp base. The bulb housing is also modifiable onthe lamp arm. It is sold individually.
Catalog No. NC9423184 Supplier Andwin Scientific Supplier No. 36547009
The Infrared Heating and Drying Lamp gives onvenient and dependable source for near-infrared rays for heating and drying purposes. The arm is offset in order to permit the evaporating dish or other object to be placed and heated directly under the lamp. The switch to turn it on or off is located on the lamp base. The bulb housing is also modifiable onthe lamp arm. It is sold individually.