Learn More
Research Products International Corp Infrared Laser Thermometer, Provides Instant Temperature Measurement
SDP
Catalog No. 50476589
Encompass
IR Infrared Thermometer can quickly and accurately determine the (surface) temperature of a target by measuring the infrared energy radiated by the surface. The 12:1 distance to spot ratio is suitable for non-contact temperature measurement, and has a range of -58°F ~ +932°F. Ideal for measuring  liquid temperatures without contacting solution. Provides instant temperature measurement. Simply aim the laser guide at the target and press the trigger to read temperature in °F or °C. Displays temperature within a second and prevents cross contamination between samples. Other functions include temperature alarm (high/ low) and auto power off (15 seconds).  Supplied with 2 x AAA batteries.
Fast reading
Wide temperature range
Easy operation
1 Year warranty
Catalog No. 50-476-589 Supplier Research Products International Corp Supplier No. 800047
IR Infrared Thermometer can quickly and accurately determine the (surface) temperature of a target by measuring the infrared energy radiated by the surface. The 12:1 distance to spot ratio is suitable for non-contact temperature measurement, and has a range of -58°F ~ +932°F. Ideal for measuring  liquid temperatures without contacting solution. Provides instant temperature measurement. Simply aim the laser guide at the target and press the trigger to read temperature in °F or °C. Displays temperature within a second and prevents cross contamination between samples. Other functions include temperature alarm (high/ low) and auto power off (15 seconds).  Supplied with 2 x AAA batteries.
Fast reading
Wide temperature range
Easy operation
1 Year warranty