Learn More
Stoelting™ Infrared Night Light Illuminator
SDP
Catalog No. 10000335 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-335 Neuroscience
1 options
Catalog No. 10-000-335 Supplier Stoelting™ Supplier No. 60540
Add to Cart
Add to Cart
Stoelting Infrared Night Light Illuminator (IR illuminators) is typically used when ambient light is either not present or too dim.The IR illuminator is a source of invisible infrared light to provide night vision.
Specifications
For Use With (Application) Neuroscience