Learn More
SAI Infusion Technologies 45" Extension Line - PU, with 23G Catheter Connector, 25/pk
SDP
Catalog No. NC1403145
Encompass
45" PU Extension Line with a 23G stainless steel catheter connector. Open end is accessible with a 23G blunt needle (such as B23-50, sold separately).These extension lines are sterile, 25/pk. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC1403145 Supplier SAI Infusion Technologies Supplier No. EXTP4523CC
45" PU Extension Line with a 23G stainless steel catheter connector. Open end is accessible with a 23G blunt needle (such as B23-50, sold separately).These extension lines are sterile, 25/pk. Product is for laboratory animal research only.