Learn More
Med Vet International Infusion Pipettes, Drilled, J0287B, 18in, 25 Count
Supplier:  Med Vet International JOR287B
Encompass_Preferred
High quality PVC pipette with rounded polished insertion end to prevent tissue trauma. Large bore, manufactured with the following specifications; 0.160 i.d. x 0.225 o.d. for infusing viscous fluids. All tubes 25/poly bag.
Catalog No. 50-005-82981