Learn More
Med Vet International Infusion Pipettes, Drilled, Sterile, J0287E, 21in, 25 Count
Catalog No. 5000581610
Encompass_Preferred
High-quality PVC pipette with rounded, polished insertion end to prevent tissue trauma. Large bore, manufactured with the following specifications: 0.160 i.d. x 0.225 o.d. for infusing viscous fluids. All tubes 25/poly bag.
Catalog No. 50-005-81610 Supplier Med Vet International Supplier No. JOR287E
High-quality PVC pipette with rounded, polished insertion end to prevent tissue trauma. Large bore, manufactured with the following specifications: 0.160 i.d. x 0.225 o.d. for infusing viscous fluids. All tubes 25/poly bag.