Learn More
Med Vet International Infusion Pipettes, Flex Adaptor, J0287A, 18in, 25 Count
Catalog No. 5000582608
Encompass_Preferred
High quality PVC pipette with rounded polished insertion end to prevent tissue trauma. Large bore, manufactured with the following specifications; 0.160 i.d. x 0.225 o.d. for infusing viscous fluids. All tubes 25/poly bag.
Catalog No. 50-005-82608 Supplier Med Vet International Supplier No. JOR287A
Add to Cart
Add to Cart
High quality PVC pipette with rounded polished insertion end to prevent tissue trauma. Large bore, manufactured with the following specifications; 0.160 i.d. x 0.225 o.d. for infusing viscous fluids. All tubes 25/poly bag.