Learn More
Med Vet International Infusion Pipettes, Flex Adaptor, J0287F, 21in, 25 Count
Catalog No. 5000582088
Encompass_Preferred
High quality PVC pipette with rounded polished insertion end to prevent tissue trauma. Large bore, manufactured with the following specifications; 0.160 i.d. x 0.225 o.d. for infusing viscous fluids. All tubes 25/poly bag.
Catalog No. 50-005-82088 Supplier Med Vet International Supplier No. JOR287F
High quality PVC pipette with rounded polished insertion end to prevent tissue trauma. Large bore, manufactured with the following specifications; 0.160 i.d. x 0.225 o.d. for infusing viscous fluids. All tubes 25/poly bag.