Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Single-Syringe Infusion Pump; 115 VAC
Catalog No. NC9737805
Encompass
This features microprocessor motor control and precision gearing. Microprocessor automatically selects display units based on syringe used. Obtain precise flow rates down to 0.2 ¿L/hr. A wide dynamic range (6000:1) provides excellent flexibility. All comm
Catalog No. NC9737805 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7490000
This features microprocessor motor control and precision gearing. Microprocessor automatically selects display units based on syringe used. Obtain precise flow rates down to 0.2 ¿L/hr. A wide dynamic range (6000:1) provides excellent flexibility. All comm