Learn More
AMERICAN BIO SAFETY INFUTRACE, 2.5 GAL CUBE
SDP
Catalog No. NC2263485
Encompass_Preferred
Infutrace A unique solution designed for use as a secondary perfusion solution, point injection solution, a specimen dip or a spray solution for human/animal cadavers, tissue-parts, organs, or pro-sections. InfuTrace™ gently complexes formaldehyde, destroying its harmful vapors without compromising tissue fixation. It significantly reduces the risk of formaldehyde vapor exposure to faculty and students in anatomy and biology labs.
Catalog No. NC2263485 Supplier AMERICAN BIO SAFETY Supplier No. IT250
Infutrace A unique solution designed for use as a secondary perfusion solution, point injection solution, a specimen dip or a spray solution for human/animal cadavers, tissue-parts, organs, or pro-sections. InfuTrace™ gently complexes formaldehyde, destroying its harmful vapors without compromising tissue fixation. It significantly reduces the risk of formaldehyde vapor exposure to faculty and students in anatomy and biology labs.